Learn More
Aviva Systems Biology Corp IFIT3 Antibody - N-terminal region (ARP46034_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46034_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFIT3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human IFIT3. Alternate names: CIG-49, GARG-49, IFI60, IFIT4, IRG2, ISG60, RIG-GApplicationIHC, WB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59187