Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci IFIT3 Peptide
SDP

Catalog No. 16596361
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-596-361 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-596-361 Supplier ProSci Supplier No. 7759P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

The peptide is used for blocking the activity of IFIT3 Antibody (catalog number 7759). It usually blocks the antibody activity completely by incubating the peptide with equal volume of antibody for 30 min at 37°C.

Specifications

Gene ID (Entrez) 3437
Species Human
Accession Number NP_001026853
Common Name interferon-induced protein with tetratricopeptide repeats 3
Concentration 200 μg/mL
Content And Storage Store IFIT3 Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol IFIT3
Quantity 0.05 mg
Sequence 18 amino acids near the carboxy terminus of human IFIT3.
Synonym P60, IRG2, IFI60, IFIT4, ISG60, RIG-G, CIG-49, GARG-49, Interferon-induced protein with tetratricopeptide repeats 3, CIG49, IFIT-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.