Learn More
Aviva Systems Biology Corp LMNTD1 Antibody - middle region (ARP53050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFLTD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IFLTD1. Alternate names: FLJ36004, Pas1c1, PAS1C1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60904