Learn More
Aat Bioquest iFluor™ 350 succinimidyl ester
SDP
Catalog No. 501952962
Encompass_Preferred
AAT Bioquest's iFluor™ dyes are optimized for labeling proteins, in particular, antibodies. These dyes are bright, photostable and have minimal quenching on proteins.
Catalog No. 50-195-2962 Supplier Aat Bioquest Supplier No. 1020
AAT Bioquest's iFluor™ dyes are optimized for labeling proteins, in particular, antibodies. These dyes are bright, photostable and have minimal quenching on proteins.