Learn More
Aat Bioquest iFluor 546 goat anti-rabbit IgG HL (200 ug)
SDP
Supplier:  Aat Bioquest 16618
Encompass_Preferred
iFluor 546 is a bright orange fluorescent dye. iFluor 546-labeled anti-IgG conjugates exhibit bright fluorescence signal and good photostability. This catalog is for 200 ug.
Catalog No. 50-224-9495