Learn More
Aat Bioquest iFluor 555 Styramide Signal Amplification (PSA™) system, 100 Slides
SDP
Catalog No. 502250200
Encompass_Preferred
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents. In combination with our superior iFluor dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC. This catalog is for 100 slides.
This item is not returnable. View return policy
Catalog No. 50-225-0200 Supplier Aat Bioquest Supplier No. 45027
Add to Cart
Add to Cart
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents. In combination with our superior iFluor dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC. This catalog is for 100 slides.