Learn More
Aat Bioquest iFluor 647 Styramide Superior Replacement for Alexa Fluor 647 tyramide (100 Slides)
SDP
Catalog No. 502250201
Encompass_Preferred
Power Styramide Signal Amplification PSA system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide TSA reagents. This catalog is for 100 Slides.
This item is not returnable. View return policy
Catalog No. 50-225-0201 Supplier Aat Bioquest Supplier No. 45045
Power Styramide Signal Amplification PSA system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide TSA reagents. This catalog is for 100 Slides.