Learn More
Abcam iFluor Red 750 Conjugation Kit, 100UG
Catalog No. 502700607
Encompass_Preferred
iFluor Red 750 conjugation / labeling in < 4 hrs with 30 secs hands-on time using iFluor Red 750 Conjugation Kit ab312811. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-0607 Supplier Abcam Supplier No. AB312811100UG
iFluor Red 750 conjugation / labeling in < 4 hrs with 30 secs hands-on time using iFluor Red 750 Conjugation Kit ab312811. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement