Learn More
Abcam iFluor Red 750 Conjugation Kit (BSA compatible), 100UG
Catalog No. 502700606
Encompass_Preferred
iFluor Red 750 conjugation / labeling in < 4 hrs with 30 secs hands-on time using iFluor Red 750 Conjugation Kit (BSA compatible) ab312812. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-0606 Supplier Abcam Supplier No. AB312812100UG
iFluor Red 750 conjugation / labeling in < 4 hrs with 30 secs hands-on time using iFluor Red 750 Conjugation Kit (BSA compatible) ab312812. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement