Learn More
Aat Bioquest iFluor™ 488 amine
SDP
Catalog No. 501952987
Encompass_Preferred
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
Catalog No. 50-195-2987 Supplier Aat Bioquest Supplier No. 1072
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.