Learn More
Aat Bioquest iFluor™ 750 amine
SDP
Catalog No. 501952994
Encompass_Preferred
AAT Bioquest's iFluor™ dyes are developed for labeling proteins, in particular, antibodies. These dyes are optimized to have minimal fluorescence quenching effect on proteins and nucleic acids.
Catalog No. 50-195-2994 Supplier Aat Bioquest Supplier No. 1079
AAT Bioquest's iFluor™ dyes are developed for labeling proteins, in particular, antibodies. These dyes are optimized to have minimal fluorescence quenching effect on proteins and nucleic acids.