Learn More
Abcam iFluor Ultraviolet 350 Conjugation Kit, 100UG
Catalog No. 502669547
Encompass_Preferred
iFluor™ 350 conjugation / labeling in < 4 hrs with 30 secs hands-on time using iFluor Ultraviolet 350 Conjugation Kit ab312809. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-9547 Supplier Abcam Supplier No. AB312809100UG
iFluor™ 350 conjugation / labeling in < 4 hrs with 30 secs hands-on time using iFluor Ultraviolet 350 Conjugation Kit ab312809. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement