Learn More
Enzo Life Sciences IFN-alpha-2b (human), (recombinant) (active) (10µg)
SDP
Catalog No. 502013980
Encompass_Preferred
Alternative name: Interferon alpha 2b. Concentration: 0.1 mg/ml when reconstituted as suggested. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. In PBS. Source: Produced in HEK cells. The human cytokine (aa 24-188) is fused at the N-terminus to a FLAG-tag followed by an 8 AA linker. Long Term Storage: -20°C.
Catalog No. 50-201-3980 Supplier Enzo Life Sciences Supplier No. ENZPRT1925010
Alternative name: Interferon alpha 2b. Concentration: 0.1 mg/ml when reconstituted as suggested. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. In PBS. Source: Produced in HEK cells. The human cytokine (aa 24-188) is fused at the N-terminus to a FLAG-tag followed by an 8 AA linker. Long Term Storage: -20°C.