Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0300350
Encompass
Interferon-Alpha 2a (IFN-Alpha 2a) Human produced by leukocytes is a member of Interferon family. IFN-alpha is mainly involved in innate immune response against a broad range of viral infections. IFN-alpha 2 has three acid stable forms (a b c) signaling through IFNAR2. IFN-alpha 2a shares 99.4% 98.8% aa sequence identity with IFN-alpha 2b and 2c respectively. IFN-alpha contains four highly conserved cysteine residues which form two disulfide bonds one of which is necessary for biological activity.
Catalog No. 50-252-9898