Learn More
ACROBiosystems Cynomolgus IFN-alpha/beta R1 Protein, His Tag (MALS verified)
Catalog No. 502719371
Encompass_Preferred
The protein encoded by this gene is a type I membrane protein that forms one of the two chains of a receptor for interferons alpha and beta. Binding and activation of the receptor stimulates Janus protein kinases, which in turn phosphorylate several proteins, including STAT1 and STAT2. The protein belongs to the type II cytokine receptor family and functions as an antiviral factor.
Catalog No. 50-271-9371 Supplier ACROBiosystems Supplier No. IF1C52H31MG
The protein encoded by this gene is a type I membrane protein that forms one of the two chains of a receptor for interferons alpha and beta. Binding and activation of the receptor stimulates Janus protein kinases, which in turn phosphorylate several proteins, including STAT1 and STAT2. The protein belongs to the type II cytokine receptor family and functions as an antiviral factor.