Learn More
Medchemexpress LLC IFN-alpha/beta R2 protein, human (HEK293, His, solution) | 10 UG
SDP
Catalog No. 5000291626
Encompass_Preferred
Recombinant human interferon alpha/beta receptor 2 (IFNAR2) produced in HEK293 cells and supplied as an aqueous solution with a C-terminal His tag. Intended for research use in receptor binding, signaling, and protein-protein interaction studies. Molecular weight is approximately 45 kDa; consult the product Certificate of Analysis for concentration and storage instructions.
  • Human sequence expressed in HEK293 cells for mammalian-like glycosylation.
  • C-terminal His tag facilitates affinity purification and detection.
  • Supplied as a solution for immediate use in biochemical assays.
  • Suitable for receptor binding and signaling studies.
  • Accompanied by an SDS; consult the Certificate of Analysis for lot-specific data.
Catalog No. 50-002-91626 Supplier Medchemexpress LLC Supplier No. HYP74862A10UG
Recombinant human interferon alpha/beta receptor 2 (IFNAR2) produced in HEK293 cells and supplied as an aqueous solution with a C-terminal His tag. Intended for research use in receptor binding, signaling, and protein-protein interaction studies. Molecular weight is approximately 45 kDa; consult the product Certificate of Analysis for concentration and storage instructions.
  • Human sequence expressed in HEK293 cells for mammalian-like glycosylation.
  • C-terminal His tag facilitates affinity purification and detection.
  • Supplied as a solution for immediate use in biochemical assays.
  • Suitable for receptor binding and signaling studies.
  • Accompanied by an SDS; consult the Certificate of Analysis for lot-specific data.