Learn More
Medchemexpress LLC IFN-alpha/beta R2 protein, human (HEK293, His, solution) | 2 UG
SDP
Catalog No. 5000291186
Encompass_Preferred
Recombinant human interferon alpha/beta receptor 2 (IFNAR2) produced in HEK293 cells and purified with a polyhistidine tag. Supplied as a ready-to-use solution for research applications involving type I interferon signaling, receptor binding studies, and biochemical characterization.
  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Supplied as a solution for convenient use in assays.
  • Approximate molecular weight 45 kDa.
  • Suitable for receptor interaction and signaling studies.
  • Stored under recommended conditions to preserve activity.
Catalog No. 50-002-91186 Supplier Medchemexpress LLC Supplier No. HYP74862A2UG
Recombinant human interferon alpha/beta receptor 2 (IFNAR2) produced in HEK293 cells and purified with a polyhistidine tag. Supplied as a ready-to-use solution for research applications involving type I interferon signaling, receptor binding studies, and biochemical characterization.
  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Supplied as a solution for convenient use in assays.
  • Approximate molecular weight 45 kDa.
  • Suitable for receptor interaction and signaling studies.
  • Stored under recommended conditions to preserve activity.