Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IFN alpha ProQuantum Immunoassay Kit
GREENER_CHOICE

Catalog No. A42897
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42897 96 Tests
A428975 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A42897 Supplier Invitrogen™ Supplier No. A42897
Add to Cart
Add to Cart

ELISA

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01562, P01563
Assay Range 0.064-5000 pg/mL
Assay Sensitivity <0.05 pg/mL
Conjugate FAM
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Protein Quantitation
For Use With (Equipment) qPCR Instruments
Gene Alias alpha-2a interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN alpha 1/13; IFN alpha 1b; IFN α; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNα; INFA2; interferon; interferon alpha; interferon alpha 1; Interferon alpha 13 (IFNA13); interferon alpha 15; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; Interferon alpha leukocyte (IFL); interferon alpha protein; interferon alpha-1; Interferon alpha1 (IFNA1); interferon alpha-1 precursor; interferon alpha-1/13; Interferon alpha-1/13 (IFN-alpha-1/13); Interferon alpha-2; Interferon alpha-A; Interferon alpha-D; Interferon alpha-D (IFN alphaD); interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655alpha-2a interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN alpha 1/13; IFN alpha 1b; IFN α; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNα; INFA2; interferon; interferon alpha; interferon alpha 1; Interferon alpha 13 (IFNA13); interferon alpha 15; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; Interferon alpha leukocyte (IFL); interferon alpha protein; interferon alpha-1; Interferon alpha1 (IFNA1); interferon alpha-1 precursor; interferon alpha-1/13; Interferon alpha-1/13 (IFN-alpha-1/13); Interferon alpha-2; Interferon alpha-A; Interferon alpha-D; Interferon alpha-D (IFN alphaD); interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655;
Gene ID (Entrez) 3439, 3440
Gene Symbol IFNA1, IFNA2
Incubation Time 1 hr.
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Shipping Condition Dry Ice
Storage Requirements -20°C
Target IFN alpha
Test Time 30 min
Total Assay Time 2 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.