Learn More
Selleck Chemical LLC IFN alpha-IFNAR-IN-1 HCl E2982-5MG
SDP
Catalog No. 5000040765
Encompass_Preferred
IFN alpha-IFNAR-IN-1 hydrochloride is a nonpeptidic low-molecular-weight inhibitor that inhibits the interaction between IFN- and IFNAR IFN alpha-IFNAR-IN-1 hydrochloride inhibits modified Vaccinia virus ankara (MVA)-induced IFN- responses in murine bone-marrow-derived Flt3- L-differentiated pDC cultures (BM-pDCs) with IC50 value of 2-8 M
Catalog No. 50-000-40765 Supplier Selleck Chemical LLC Supplier No. E29825MG
IFN alpha-IFNAR-IN-1 hydrochloride is a nonpeptidic low-molecular-weight inhibitor that inhibits the interaction between IFN- and IFNAR IFN alpha-IFNAR-IN-1 hydrochloride inhibits modified Vaccinia virus ankara (MVA)-induced IFN- responses in murine bone-marrow-derived Flt3- L-differentiated pDC cultures (BM-pDCs) with IC50 value of 2-8 M