Learn More
Genscript Corporation Antibody Protein Proteins Proteins 5 UG 1/EA
Supplier:  Genscript Corporation Z031095
Encompass
Interferon-beta (IFN-beta ) acting via STAT1 and STAT2 is known to upregulate and downregulate a wide variety of genes most of which are involved in the antiviral immune response. It is a member of Type I IFNs which include IFN-Alpha -beta τ and –ω. IFN-beta plays an important role in inducing non-specific resistance against a broad range of viral infections. It also affects cell proliferation and modulates immune responses.
Catalog No. 50-252-9831