Learn More
Medchemexpress LLC IFN-beta Human HEK 50ug
SDP
Catalog No. 5000180201
Encompass_Preferred
IFN- (interferon- ) is a key type I interferon cytokine that coordinates the innate immune response to infection tumors and inflammation IFN- binds to high-affinity (IFNAR2) and low-affinity (IFNAR1) heterodimeric receptors activates Jak-STAT signaling and modulates interferon-regulated genes IFN-beta Protein Human (HEK293 Fc) is the recombinant human-derived IFN-beta protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80201 Supplier Medchemexpress LLC Supplier No. HYP7312950UG
IFN- (interferon- ) is a key type I interferon cytokine that coordinates the innate immune response to infection tumors and inflammation IFN- binds to high-affinity (IFNAR2) and low-affinity (IFNAR1) heterodimeric receptors activates Jak-STAT signaling and modulates interferon-regulated genes IFN-beta Protein Human (HEK293 Fc) is the recombinant human-derived IFN-beta protein expressed by HEK293 with C-hFc labeled tag