Learn More
Medchemexpress LLC IFN-beta Mouse HEK 10ug
SDP
Catalog No. 5000180460
Encompass_Preferred
IFN-beta is a secretory cytokine produced by cells under viral infection or nucleic acid stimulation IFN binds to cell surface heterodimeric receptors (IFNAR1/IFNAR2) activates the JAK-STAT signaling pathway and induces the expression of interferon-stimulated genes (ISGs) (such as PKR 2 5 -OAS) IFN-beta inhibits viral proliferation regulates immune responses (such as inhibiting Th1 cell polarization reducing pro-inflammatory cytokine secretion) inhibits the proliferation of vascular smooth muscle cells and tumor cells IFN-beta has anti-atherosclerotic and vascular remodeling effects Mouse (HEK293) is the recombinant mouse-derived IFN-beta protein expressed by HEK293 with tag free
Catalog No. 50-001-80460 Supplier Medchemexpress LLC Supplier No. HYP7313010UG
IFN-beta is a secretory cytokine produced by cells under viral infection or nucleic acid stimulation IFN binds to cell surface heterodimeric receptors (IFNAR1/IFNAR2) activates the JAK-STAT signaling pathway and induces the expression of interferon-stimulated genes (ISGs) (such as PKR 2 5 -OAS) IFN-beta inhibits viral proliferation regulates immune responses (such as inhibiting Th1 cell polarization reducing pro-inflammatory cytokine secretion) inhibits the proliferation of vascular smooth muscle cells and tumor cells IFN-beta has anti-atherosclerotic and vascular remodeling effects Mouse (HEK293) is the recombinant mouse-derived IFN-beta protein expressed by HEK293 with tag free