Learn More
Medchemexpress LLC IFN-beta Protein Mo 2ug
SDP
Catalog No. 5000180481
Encompass_Preferred
IFN- protein is a type I interferon that plays a key role in the innate immune response by binding to IFNAR2 and IFNAR1 receptors and activating Jak-STAT signaling This results in transcriptional regulation of interferon-regulated genes affecting antiviral proteins cell proliferation regulators and immunomodulatory proteins IFN-beta Protein Mouse (HEK293 Fc) is the recombinant mouse-derived IFN-beta protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80481 Supplier Medchemexpress LLC Supplier No. HYP731312UG
IFN- protein is a type I interferon that plays a key role in the innate immune response by binding to IFNAR2 and IFNAR1 receptors and activating Jak-STAT signaling This results in transcriptional regulation of interferon-regulated genes affecting antiviral proteins cell proliferation regulators and immunomodulatory proteins IFN-beta Protein Mouse (HEK293 Fc) is the recombinant mouse-derived IFN-beta protein expressed by HEK293 with C-hFc labeled tag