Learn More
Medchemexpress LLC IFN-beta Rhesus Mac 10ug
SDP
Catalog No. 5000181061
Encompass_Preferred
IFN-beta Protein is an immunomodulatory cytokine of the type 1 interferon family IFN-beta Protein reduces joint inflammation by inhibiting the RANKL-c-Fos signaling pathway IFN-beta Protein regulates mitochondrial fission through STAT5 PGAM5 and Drp1 to rescue neurodegeneration IFN-beta Protein Rhesus Macaque (HEK293 Fc) is the recombinant Rhesus Macaque-derived IFN-beta protein expressed by HEK293 with C-mFc labeled tag
Catalog No. 50-001-81061 Supplier Medchemexpress LLC Supplier No. HYP7313210UG
IFN-beta Protein is an immunomodulatory cytokine of the type 1 interferon family IFN-beta Protein reduces joint inflammation by inhibiting the RANKL-c-Fos signaling pathway IFN-beta Protein regulates mitochondrial fission through STAT5 PGAM5 and Drp1 to rescue neurodegeneration IFN-beta Protein Rhesus Macaque (HEK293 Fc) is the recombinant Rhesus Macaque-derived IFN-beta protein expressed by HEK293 with C-mFc labeled tag