Learn More
Medchemexpress LLC IFN-epsilon Human 10ug | 10ug
SDP
Catalog No. 5000196779
Encompass_Preferred
IFN- protein is a type I interferon that is critical for maintaining baseline levels of IFN-regulated genes (such as 2 -5 -oligoadenylate synthetase IRF7 and ISG15) in the female reproductive tract It acts as a direct mediator providing protection against viral and bacterial genital infections IFN-epsilon Protein Human (His) is the recombinant human-derived IFN-epsilon protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-96779 Supplier Medchemexpress LLC Supplier No. HYP7158910UG
IFN- protein is a type I interferon that is critical for maintaining baseline levels of IFN-regulated genes (such as 2 -5 -oligoadenylate synthetase IRF7 and ISG15) in the female reproductive tract It acts as a direct mediator providing protection against viral and bacterial genital infections IFN-epsilon Protein Human (His) is the recombinant human-derived IFN-epsilon protein expressed by E coli with N-6 His labeled tag