Learn More
Medchemexpress LLC IFN-gamma Canine 100ug | 100ug
SDP
Catalog No. 5000194682
Encompass_Preferred
IFN-gamma (interferon-gamma) is a type II interferon produced by immune cells such as T cells and NK cells and plays a key role in activating antibacterial antiviral and antitumor responses Through the JAK-STAT pathway its interaction with the receptor IFNGR1 triggers gene regulation IFN-gamma Protein Canine is the recombinant canine-derived IFN-gamma protein expressed by E coli with tag free
Catalog No. 50-001-94682 Supplier Medchemexpress LLC Supplier No. HYP79295100UG
IFN-gamma (interferon-gamma) is a type II interferon produced by immune cells such as T cells and NK cells and plays a key role in activating antibacterial antiviral and antitumor responses Through the JAK-STAT pathway its interaction with the receptor IFNGR1 triggers gene regulation IFN-gamma Protein Canine is the recombinant canine-derived IFN-gamma protein expressed by E coli with tag free