Learn More
Medchemexpress LLC IFN-gamma protein, human (CHO) | >95.0% | 10UG
SDP
Catalog No. 501934275
Encompass_Preferred
Recombinant human interferon-gamma expressed in CHO cells, supplied as a tag-free, lyophilized powder for cell-based and biochemical research. The product corresponds to mature human IFN-γ (residues Q24-Q166, accession P01579), with reported biological activity and suitability for antiviral and immunology assays.
  • Produced in CHO cells to preserve mammalian post-translational modifications.
  • Tag free, lyophilized formulation for flexible reconstitution.
  • >95.0% purity by reducing SDS-PAGE.
  • Reported specific activity in cell-based assays, suitable for functional studies.
  • Approximate molecular mass 15-25 kDa due to glycosylation.
  • Stable when stored lyophilized at -20°C; aliquot and freeze at -80°C after reconstitution.
Catalog No. 50-193-4275 Supplier Medchemexpress LLC Supplier No. HYP7025A10UG
Add to Cart
Add to Cart
Recombinant human interferon-gamma expressed in CHO cells, supplied as a tag-free, lyophilized powder for cell-based and biochemical research. The product corresponds to mature human IFN-γ (residues Q24-Q166, accession P01579), with reported biological activity and suitability for antiviral and immunology assays.
  • Produced in CHO cells to preserve mammalian post-translational modifications.
  • Tag free, lyophilized formulation for flexible reconstitution.
  • >95.0% purity by reducing SDS-PAGE.
  • Reported specific activity in cell-based assays, suitable for functional studies.
  • Approximate molecular mass 15-25 kDa due to glycosylation.
  • Stable when stored lyophilized at -20°C; aliquot and freeze at -80°C after reconstitution.