Learn More
Medchemexpress LLC IFN-gamma Feline 10ug | 10ug
SDP
Catalog No. 5000194729
Encompass_Preferred
IFN-gamma (Interferon-gamma) is a type II interferon produced by immune cells such as T cells and NK cells which can significantly activate antibacterial antiviral and anti-tumor responses Its main JAK-STAT signaling pathway is triggered by interaction with the receptor IFNGR1 affecting gene regulation IFN-gamma Protein Feline is the recombinant IFN-gamma protein expressed by E coli with tag free
Catalog No. 50-001-94729 Supplier Medchemexpress LLC Supplier No. HYP7928010UG
IFN-gamma (Interferon-gamma) is a type II interferon produced by immune cells such as T cells and NK cells which can significantly activate antibacterial antiviral and anti-tumor responses Its main JAK-STAT signaling pathway is triggered by interaction with the receptor IFNGR1 affecting gene regulation IFN-gamma Protein Feline is the recombinant IFN-gamma protein expressed by E coli with tag free