Learn More
Enzo Life Sciences IFN-gamma (human), (recombinant) (100µg)
SDP
Catalog No. 502013932
Encompass_Preferred
Alternative name: Interferon-gamma. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10mM sodium phosphate, pH 7.4. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3932 Supplier Enzo Life Sciences Supplier No. ENZPRT1410100
Alternative name: Interferon-gamma. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10mM sodium phosphate, pH 7.4. Source: Produced in E.coli. Long Term Storage: -20°C.