Learn More
Medchemexpress LLC HY-P7025 10ug Medchemexpress, IFN-gamma, Human CAS: Purity:>98%
SDP
Catalog No. 501970556
Encompass_Preferred
Medchemexpress, HY-P7025 10ug IFN-gamma, Human CAS: Recombinant Human Interferon-gamma is a cytokine with potent immunomodulatory, antiviral and antitumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0556 Supplier Medchemexpress LLC Supplier No. HYP702510UG
Medchemexpress, HY-P7025 10ug IFN-gamma, Human CAS: Recombinant Human Interferon-gamma is a cytokine with potent immunomodulatory, antiviral and antitumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.