Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon gamma protein, human | 82115-62-6 | >95.0% | 1MG
SDP

Catalog No. 5000175597
Encompass_Preferred

Recombinant human interferon-gamma (IFN-γ) supplied as a lyophilized purified protein for research use. The product corresponds to human IFN-γ (UniProt P01579, residues Q24-Q166), produced in Escherichia coli, with a predicted molecular mass of ~16.7 kDa and purity greater than 95% by reducing SDS-PAGE. Intended for immunology, cell signaling, antiviral, and antitumor research; store lyophilized at -20°C and aliquot for long-term storage.

  • Recombinant human interferon-gamma (residues Q24-Q166)
  • Produced in Escherichia coli expression system
  • Lyophilized formulation for extended shelf life
  • Purity greater than 95% by reducing SDS-PAGE
  • Predicted molecular weight ~16.7 kDa
  • Suitable for STAT signaling, antiviral, and immunomodulation assays
  • Stable when aliquoted and stored at -20°C to -80°C

Catalog No. 50-001-75597 Supplier Medchemexpress LLC Supplier No. HYP70251MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon-gamma (IFN-γ) supplied as a lyophilized purified protein for research use. The product corresponds to human IFN-γ (UniProt P01579, residues Q24-Q166), produced in Escherichia coli, with a predicted molecular mass of ~16.7 kDa and purity greater than 95% by reducing SDS-PAGE. Intended for immunology, cell signaling, antiviral, and antitumor research; store lyophilized at -20°C and aliquot for long-term storage.

  • Recombinant human interferon-gamma (residues Q24-Q166)
  • Produced in Escherichia coli expression system
  • Lyophilized formulation for extended shelf life
  • Purity greater than 95% by reducing SDS-PAGE
  • Predicted molecular weight ~16.7 kDa
  • Suitable for STAT signaling, antiviral, and immunomodulation assays
  • Stable when aliquoted and stored at -20°C to -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.