Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon-gamma, human recombinant | 98059-61-1 | 2UG
SDP

Catalog No. 5000175353
Encompass_Preferred

Recombinant human interferon-gamma is a cytokine used to activate immune effector cells, enhance antigen presentation, and study IFN-γ-mediated signaling pathways in vitro. Supplied as a lyophilized protein with verified bioactivity and low endotoxin content, it is suitable for cell culture stimulation, signaling assays, and immunology research.

  • Recombinant human IFN-gamma (Gln24-Gln166).
  • Potent immunomodulatory and antiviral activity.
  • High specific activity (ED50 <1 ng/mL).
  • Low endotoxin level (<1 EU/μg).
  • Lyophilized formulation for improved stability.
  • Expressed in E. coli; molecular weight ~16-18 kDa.
  • Store at -20°C; aliquot and freeze for extended storage.
  • Not recommended to reconstitute below 100 μg/mL.

Catalog No. 50-001-75353 Supplier Medchemexpress LLC Supplier No. HYP70252UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon-gamma is a cytokine used to activate immune effector cells, enhance antigen presentation, and study IFN-γ-mediated signaling pathways in vitro. Supplied as a lyophilized protein with verified bioactivity and low endotoxin content, it is suitable for cell culture stimulation, signaling assays, and immunology research.

  • Recombinant human IFN-gamma (Gln24-Gln166).
  • Potent immunomodulatory and antiviral activity.
  • High specific activity (ED50 <1 ng/mL).
  • Low endotoxin level (<1 EU/μg).
  • Lyophilized formulation for improved stability.
  • Expressed in E. coli; molecular weight ~16-18 kDa.
  • Store at -20°C; aliquot and freeze for extended storage.
  • Not recommended to reconstitute below 100 μg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.