Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human interferon gamma | 98059-61-1 | >95.0% | 500UG
SDP

Catalog No. 5000175864
Encompass_Preferred

Recombinant human interferon-gamma (IFN-γ) is a biologically active cytokine provided as a lyophilized protein for immunology and cell-signaling research. It corresponds to the mature human IFN-γ sequence (Q24-Q166), is expressed in E. coli, and is characterized for purity, activity, and low endotoxin for reliable experimental use.

  • Biologically active recombinant human IFN-γ for cell signaling and immunology assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Specific activity >1 x 10^6 units/mg; ED50 <1 ng/mL (HT-29 assay).
  • Low endotoxin level: <1 EU/μg (LAL).
  • Lyophilized formulation with stabilizers for improved shelf life.
  • Expressed in E. coli; mature sequence Q24-Q166.
  • Predicted mass ~16.7 kDa (≈16-18 kDa by SDS-PAGE).
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen for long-term storage.

Catalog No. 50-001-75864 Supplier Medchemexpress LLC Supplier No. HYP7025500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon-gamma (IFN-γ) is a biologically active cytokine provided as a lyophilized protein for immunology and cell-signaling research. It corresponds to the mature human IFN-γ sequence (Q24-Q166), is expressed in E. coli, and is characterized for purity, activity, and low endotoxin for reliable experimental use.

  • Biologically active recombinant human IFN-γ for cell signaling and immunology assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Specific activity >1 x 10^6 units/mg; ED50 <1 ng/mL (HT-29 assay).
  • Low endotoxin level: <1 EU/μg (LAL).
  • Lyophilized formulation with stabilizers for improved shelf life.
  • Expressed in E. coli; mature sequence Q24-Q166.
  • Predicted mass ~16.7 kDa (≈16-18 kDa by SDS-PAGE).
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.