Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IFN-gamma protein, human (CHO) | >95.0% | 100UG
SDP

Catalog No. 5000176103
Encompass_Preferred

IFN-gamma Protein, Human (CHO) is a recombinant human cytokine produced in Chinese hamster ovary (CHO) cells and supplied as a lyophilized, tag-free protein for research use. It is intended for studies of immune signaling, antiviral responses, and cell-based bioassays, and is provided with supporting documentation including a safety data sheet and certificate of analysis.

  • Recombinant human interferon-gamma expressed in CHO cells.
  • Tag free, lyophilized formulation with trehalose stabilizer.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~15-25 kDa due to glycosylation.
  • Characterized bioactivity with low-nanogram ED50 in standard assays.
  • Stable when stored at -20°C; aliquot and freeze at -80°C for long-term storage.
  • Includes SDS and COA documentation for quality and safety.

Catalog No. 50-001-76103 Supplier Medchemexpress LLC Supplier No. HYP7025A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IFN-gamma Protein, Human (CHO) is a recombinant human cytokine produced in Chinese hamster ovary (CHO) cells and supplied as a lyophilized, tag-free protein for research use. It is intended for studies of immune signaling, antiviral responses, and cell-based bioassays, and is provided with supporting documentation including a safety data sheet and certificate of analysis.

  • Recombinant human interferon-gamma expressed in CHO cells.
  • Tag free, lyophilized formulation with trehalose stabilizer.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~15-25 kDa due to glycosylation.
  • Characterized bioactivity with low-nanogram ED50 in standard assays.
  • Stable when stored at -20°C; aliquot and freeze at -80°C for long-term storage.
  • Includes SDS and COA documentation for quality and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.