Learn More
Enzo Life Sciences IFN-gamma (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900136
Encompass_Preferred
The IFN-γ (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Alternative Name: Interferon-gamma. Sensitivity: < 2 pg/ml (range 25.6 - 1,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, Biotinylated antibody, Standard, Standard diluent, Wash buffer concentrate, TMB Substrate, Stop solution
Catalog No. 50-658-166