Recombinant human interferon-gamma (IFN-γ) produced in HEK293 cells and supplied without an affinity tag for research applications. The protein corresponds to UniProt accession P01579 (Q24-Q166), is >95% pure by reducing SDS-PAGE, and demonstrates potent biological activity in cell-based proliferation assays. Store frozen at -20°C; after reconstitution it is stable short-term at 4°C and for long-term storage at -20°C or -80°C when aliquoted.
- Recombinant human protein expressed in HEK293 cells.
- Tag free to minimize experimental interference.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- High specific activity; ED50 in HT-29 proliferation assay ≤0.4622 ng/mL.
- Supplied in multiple research sizes including 10 μg.
- Store at -20°C; aliquot and avoid repeated freeze-thaw cycles.
- Suitable for studies of signaling, immune modulation, and cytokine function.