Learn More
Medchemexpress LLC IFN-gamma Human HE 2ug
SDP
Catalog No. 5000184202
Encompass_Preferred
Recombinant human interferon-gamma expressed in HEK293 cells and supplied lyophilized for research use. The protein is characterized for purity, molecular mass, formulation, storage stability, and biological activity, making it suitable for cell-based assays and immunological studies.
  • Produced in HEK293 mammalian expression system.
  • Lyophilized formulation for long-term storage and transport.
  • High purity, >95% by reducing SDS-PAGE.
  • Characterized biological activity with low ED50 and high specific activity.
  • Includes recommended reconstitution, aliquoting, and storage instructions.
  • Sequence corresponds to the human IFN-γ mature form (UniProt P01579).
Catalog No. 50-001-84202 Supplier Medchemexpress LLC Supplier No. HYP706102UG
Recombinant human interferon-gamma expressed in HEK293 cells and supplied lyophilized for research use. The protein is characterized for purity, molecular mass, formulation, storage stability, and biological activity, making it suitable for cell-based assays and immunological studies.
  • Produced in HEK293 mammalian expression system.
  • Lyophilized formulation for long-term storage and transport.
  • High purity, >95% by reducing SDS-PAGE.
  • Characterized biological activity with low ED50 and high specific activity.
  • Includes recommended reconstitution, aliquoting, and storage instructions.
  • Sequence corresponds to the human IFN-γ mature form (UniProt P01579).