Learn More
Medchemexpress LLC IFN-gamma Human HE 50ug | 50ug
SDP
Catalog No. 5000194431
Encompass_Preferred
IFN-gamma Protein is a dimeric soluble cytokine It exerts antibacterial antiviral and antitumor effects through JAK-STAT mTOR MAPK and PI3K/AKT signaling pathways IFN-gamma Protein Human (HEK293 His-Avi) is the recombinant human-derived IFN-gamma protein expressed by HEK293 with C-His C-Avi labeled tag
Catalog No. 50-001-94431 Supplier Medchemexpress LLC Supplier No. HYP7845650UG
IFN-gamma Protein is a dimeric soluble cytokine It exerts antibacterial antiviral and antitumor effects through JAK-STAT mTOR MAPK and PI3K/AKT signaling pathways IFN-gamma Protein Human (HEK293 His-Avi) is the recombinant human-derived IFN-gamma protein expressed by HEK293 with C-His C-Avi labeled tag