Learn More
Medchemexpress LLC HY-P7071 10ug Medchemexpress, IFN-gamma, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934314
Encompass_Preferred
Medchemexpress, HY-P7071 10ug IFN-gamma, Mouse CAS: Recombinant Mouse Interferon-gamma is a pro-inflammatory cytokine with potent immunomodulatory, anti-proliferative, and antiviral properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4314 Supplier Medchemexpress LLC Supplier No. HYP707110UG
Medchemexpress, HY-P7071 10ug IFN-gamma, Mouse CAS: Recombinant Mouse Interferon-gamma is a pro-inflammatory cytokine with potent immunomodulatory, anti-proliferative, and antiviral properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.