Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IFN-gamma Mouse 1mg
SDP

Catalog No. 5000184493
Encompass_Preferred

Recombinant mouse interferon-gamma (IFN-γ) is supplied as a lyophilized, bioactive protein produced in E. coli for use in immunology and cell-based research. The product offers high purity, low endotoxin, and demonstrated functional activity, making it suitable as a reagent for signaling studies, activation of immune cells, and assay controls. Store frozen and follow datasheet instructions for reconstitution and aliquoting.

  • High purity (>95%) ensures reliable assay performance.
  • Low endotoxin (<1 EU/μg) suitable for sensitive cell-based assays.
  • Verified bioactivity with ED50 ≈ 0.14 ng/mL for functional assays.
  • Sequence and accession provided for reproducibility.
  • Lyophilized format enables long-term storage and transport.
  • Recombinant expression in E. coli provides consistent lot-to-lot supply.

Catalog No. 50-001-84493 Supplier Medchemexpress LLC Supplier No. HYP70711MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interferon-gamma (IFN-γ) is supplied as a lyophilized, bioactive protein produced in E. coli for use in immunology and cell-based research. The product offers high purity, low endotoxin, and demonstrated functional activity, making it suitable as a reagent for signaling studies, activation of immune cells, and assay controls. Store frozen and follow datasheet instructions for reconstitution and aliquoting.

  • High purity (>95%) ensures reliable assay performance.
  • Low endotoxin (<1 EU/μg) suitable for sensitive cell-based assays.
  • Verified bioactivity with ED50 ≈ 0.14 ng/mL for functional assays.
  • Sequence and accession provided for reproducibility.
  • Lyophilized format enables long-term storage and transport.
  • Recombinant expression in E. coli provides consistent lot-to-lot supply.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.