Learn More
Medchemexpress LLC IFN-gamma Mouse HE 100ug
SDP
Catalog No. 5000185068
Encompass_Preferred
Recombinant mouse interferon-gamma (IFN-γ) expressed in HEK293 cells, provided as a lyophilized powder for laboratory research. The product is >95% pure by reducing SDS-PAGE, has low endotoxin (<1 EU/μg), and shows potent biological activity in cell-based assays; follow the manufacturer's reconstitution and storage recommendations for optimal performance.
  • HEK293-expressed recombinant protein suitable for in vitro studies.
  • Lyophilized formulation for long-term storage and transport.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) for sensitive assays.
  • Demonstrated biological activity with ED50 in the low ng/mL range.
  • Stable when stored frozen and after reconstitution with recommended buffers.
Catalog No. 50-001-85068 Supplier Medchemexpress LLC Supplier No. HYP70667100UG
Recombinant mouse interferon-gamma (IFN-γ) expressed in HEK293 cells, provided as a lyophilized powder for laboratory research. The product is >95% pure by reducing SDS-PAGE, has low endotoxin (<1 EU/μg), and shows potent biological activity in cell-based assays; follow the manufacturer's reconstitution and storage recommendations for optimal performance.
  • HEK293-expressed recombinant protein suitable for in vitro studies.
  • Lyophilized formulation for long-term storage and transport.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) for sensitive assays.
  • Demonstrated biological activity with ED50 in the low ng/mL range.
  • Stable when stored frozen and after reconstitution with recommended buffers.