Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IFN-gamma Mouse HE 2ug
SDP

Catalog No. 5000184297
Encompass_Preferred

Recombinant mouse interferon-gamma (IFN-γ) produced in HEK293 mammalian cells. This tag-free, glycosylated cytokine is provided as a high-purity preparation characterized by SDS-PAGE and predicted molecular mass, and is intended for in vitro immunology and cell-signaling studies such as antiviral and macrophage-activation assays.

  • Mammalian-expressed for native glycosylation.
  • Tag free to avoid interference in receptor binding assays.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass 15.5 kDa; observed ~12-21 kDa due to glycosylation.
  • Available in small research-appropriate aliquots (for example, 2 μg).
  • Includes mature amino acid sequence for reproducibility.
  • Suitable for cell signaling, immunology, and antiviral assays.

Catalog No. 50-001-84297 Supplier Medchemexpress LLC Supplier No. HYP706672UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interferon-gamma (IFN-γ) produced in HEK293 mammalian cells. This tag-free, glycosylated cytokine is provided as a high-purity preparation characterized by SDS-PAGE and predicted molecular mass, and is intended for in vitro immunology and cell-signaling studies such as antiviral and macrophage-activation assays.

  • Mammalian-expressed for native glycosylation.
  • Tag free to avoid interference in receptor binding assays.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass 15.5 kDa; observed ~12-21 kDa due to glycosylation.
  • Available in small research-appropriate aliquots (for example, 2 μg).
  • Includes mature amino acid sequence for reproducibility.
  • Suitable for cell signaling, immunology, and antiviral assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.