Learn More
Medchemexpress LLC IFN-gamma Porcine 5ug | 5ug
SDP
Catalog No. 5000194702
Encompass_Preferred
IFN-gamma (interferon-gamma) is a type II interferon produced by immune cells such as T cells and NK cells It plays a key role in antibacterial antiviral and anti-tumor responses by activating effector immune cells and enhancing antigen presentation effect Its main signaling pathway involves the JAK-STAT pathway that interacts with its receptor IFNGR1 affecting gene regulation IFN-gamma Protein Pig is the recombinant Pig-derived IFN-gamma protein expressed by E coli with tag free
Catalog No. 50-001-94702 Supplier Medchemexpress LLC Supplier No. HYP794295UG
IFN-gamma (interferon-gamma) is a type II interferon produced by immune cells such as T cells and NK cells It plays a key role in antibacterial antiviral and anti-tumor responses by activating effector immune cells and enhancing antigen presentation effect Its main signaling pathway involves the JAK-STAT pathway that interacts with its receptor IFNGR1 affecting gene regulation IFN-gamma Protein Pig is the recombinant Pig-derived IFN-gamma protein expressed by E coli with tag free