Learn More
Medchemexpress LLC IFN-gamma R1/CD119 100ug
SDP
Catalog No. 5000182601
Encompass_Preferred
IFN-gamma R1 (CD119) one of the subunit of IFN-gamma receptor is a receptor for IFN-gamma IFN-gamma R1 forms the functional receptor with IFN-gamma R2 Upon binding with IFN-gamma IFN-gamma R1 and IFN-gamma R2 oligomerize and transphosphorylate[1] Then JAK1 and JAK2 are phosphorylated and activated and STAT1 is recruited to the receptor complex Phosphorylated STAT1 translocates to the nucleus where it regulates the expression of IFN-responsive genes (e g CD54)[2] IFN-gamma R1/CD119 Protein Human (228a a HEK293 His) is a recombinant human IFN-gamma R1 (E18-G245) with C-terminal 6 His tag which is produced in HEK293 cells
Catalog No. 50-001-82601 Supplier Medchemexpress LLC Supplier No. HYP72612A100UG
IFN-gamma R1 (CD119) one of the subunit of IFN-gamma receptor is a receptor for IFN-gamma IFN-gamma R1 forms the functional receptor with IFN-gamma R2 Upon binding with IFN-gamma IFN-gamma R1 and IFN-gamma R2 oligomerize and transphosphorylate[1] Then JAK1 and JAK2 are phosphorylated and activated and STAT1 is recruited to the receptor complex Phosphorylated STAT1 translocates to the nucleus where it regulates the expression of IFN-responsive genes (e g CD54)[2] IFN-gamma R1/CD119 Protein Human (228a a HEK293 His) is a recombinant human IFN-gamma R1 (E18-G245) with C-terminal 6 His tag which is produced in HEK293 cells