Learn More
Medchemexpress LLC IFN-gamma R1/CD119 protein, rat (HEK293, Fc) | >95.0% | 500 UG
SDP
Catalog No. 5000292661
Encompass_Preferred
Recombinant rat interferon-gamma receptor 1 (IFN-γ R1/CD119) extracellular domain expressed in HEK293 cells and provided with a C-terminal Fc tag. Supplied as a sterile, 0.22 μm-filtered solution with high purity suitable for biochemical and cell-based assays, binding studies, and reagent development.
  • Recombinant rat IFN-gamma receptor 1 extracellular domain.
  • Expressed in HEK293 mammalian cells.
  • C-terminal hFc tag for dimerization and detection.
  • High purity >95.0% by reducing SDS-PAGE.
  • Supplied as 0.22 μm filtered solution in phosphate buffer.
Catalog No. 50-002-92661 Supplier Medchemexpress LLC Supplier No. HYP75824500UG
Recombinant rat interferon-gamma receptor 1 (IFN-γ R1/CD119) extracellular domain expressed in HEK293 cells and provided with a C-terminal Fc tag. Supplied as a sterile, 0.22 μm-filtered solution with high purity suitable for biochemical and cell-based assays, binding studies, and reagent development.
  • Recombinant rat IFN-gamma receptor 1 extracellular domain.
  • Expressed in HEK293 mammalian cells.
  • C-terminal hFc tag for dimerization and detection.
  • High purity >95.0% by reducing SDS-PAGE.
  • Supplied as 0.22 μm filtered solution in phosphate buffer.