Learn More
Medchemexpress LLC IFN-gamma R2 Human 10ug
SDP
Catalog No. 5000179513
Encompass_Preferred
IFN-gamma R2 one of the subunit of IFN-gamma receptor is a receptor for IFN-gamma IFN-gamma R2 is the signal-transducing chain of the IFN-gamma receptor IFN-gamma R2 forms the functional receptor with IFN-gamma R1 Upon binding with IFN-gamma IFN-gamma R2 and IFN-gamma R1 oligomerize and transphosphorylate[1] Then the downstream signaling components JAK1 and JAK2 are phosphorylated and activated and STAT1 is phosphorylated Phosphorylated STAT1 translocates to the nucleus where it regulates the expression of IFN-responsive genes (e g CD54)[2] IFN-gamma R2 Protein Human (CHO) is a recombinant human IFN-gamma R2 (S28-Q247) without any tag which is produced in CHO cells
Catalog No. 50-001-79513 Supplier Medchemexpress LLC Supplier No. HYP736710UG
IFN-gamma R2 one of the subunit of IFN-gamma receptor is a receptor for IFN-gamma IFN-gamma R2 is the signal-transducing chain of the IFN-gamma receptor IFN-gamma R2 forms the functional receptor with IFN-gamma R1 Upon binding with IFN-gamma IFN-gamma R2 and IFN-gamma R1 oligomerize and transphosphorylate[1] Then the downstream signaling components JAK1 and JAK2 are phosphorylated and activated and STAT1 is phosphorylated Phosphorylated STAT1 translocates to the nucleus where it regulates the expression of IFN-responsive genes (e g CD54)[2] IFN-gamma R2 Protein Human (CHO) is a recombinant human IFN-gamma R2 (S28-Q247) without any tag which is produced in CHO cells