Learn More
Medchemexpress LLC IFN-gamma Rat 2ug
SDP
Catalog No. 5000197092
Encompass_Preferred
Recombinant rat interferon-gamma (IFN-γ) protein supplied lyophilized for research use. This pro-inflammatory cytokine exhibits immunomodulatory, antiproliferative, and antiviral activities and is intended for use in cell signaling, immunology assays, and functional studies. The product is provided with documentation on purity, formulation, and storage to support reproducible experimental results.
  • Recombinant rat interferon-gamma protein suitable for in vitro assays.
  • Pro-inflammatory and immunomodulatory activity for functional studies.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation for extended stability and convenient storage.
  • Includes recommended reconstitution guidance to support experimental consistency.
  • Stable when stored lyophilized at -20°C; freeze aliquots after reconstitution for long-term storage.
Catalog No. 50-001-97092 Supplier Medchemexpress LLC Supplier No. HYP70952UG
Recombinant rat interferon-gamma (IFN-γ) protein supplied lyophilized for research use. This pro-inflammatory cytokine exhibits immunomodulatory, antiproliferative, and antiviral activities and is intended for use in cell signaling, immunology assays, and functional studies. The product is provided with documentation on purity, formulation, and storage to support reproducible experimental results.
  • Recombinant rat interferon-gamma protein suitable for in vitro assays.
  • Pro-inflammatory and immunomodulatory activity for functional studies.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation for extended stability and convenient storage.
  • Includes recommended reconstitution guidance to support experimental consistency.
  • Stable when stored lyophilized at -20°C; freeze aliquots after reconstitution for long-term storage.