Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IFN-gamma Rat CHO 100ug
SDP

Catalog No. 5000197157
Encompass_Preferred

IFN-gamma Protein, Rat (CHO) is a recombinant rat interferon-gamma produced in CHO cells and supplied as a lyophilized, tag-free preparation. It exhibits potent immunomodulatory, anti-proliferative, and antiviral activity and is suitable for functional assays, receptor binding studies, and cellular signaling research. The product is provided with storage and handling guidance to preserve activity.

  • Recombinant CHO-expressed rat interferon-gamma in lyophilized form.
  • Tag free to minimize interference in downstream assays.
  • High purity (>95%) for consistent experimental results.
  • Potent biological activity (ED50 <2.5 ng/mL) for sensitive functional assays.
  • Formulated with trehalose to enhance stability during storage.
  • Suitable for macrophage activation, TH1 response, and receptor binding studies.

Catalog No. 50-001-97157 Supplier Medchemexpress LLC Supplier No. HYP7095A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IFN-gamma Protein, Rat (CHO) is a recombinant rat interferon-gamma produced in CHO cells and supplied as a lyophilized, tag-free preparation. It exhibits potent immunomodulatory, anti-proliferative, and antiviral activity and is suitable for functional assays, receptor binding studies, and cellular signaling research. The product is provided with storage and handling guidance to preserve activity.

  • Recombinant CHO-expressed rat interferon-gamma in lyophilized form.
  • Tag free to minimize interference in downstream assays.
  • High purity (>95%) for consistent experimental results.
  • Potent biological activity (ED50 <2.5 ng/mL) for sensitive functional assays.
  • Formulated with trehalose to enhance stability during storage.
  • Suitable for macrophage activation, TH1 response, and receptor binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.