Learn More
Medchemexpress LLC IFN-gamma Sheep Hi 50ug
SDP
Catalog No. 5000182523
Encompass_Preferred
IFN-gamma (interferon-gamma) is a type II interferon produced by T cells and NK cells which can significantly activate antibacterial antiviral and antitumor responses It acts through the JAK-STAT pathway and its receptor IFNGR1 to affect gene regulation IFN-gamma Protein Sheep (His) is the recombinant sheep-derived IFN-gamma protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-82523 Supplier Medchemexpress LLC Supplier No. HYP7224550UG
IFN-gamma (interferon-gamma) is a type II interferon produced by T cells and NK cells which can significantly activate antibacterial antiviral and antitumor responses It acts through the JAK-STAT pathway and its receptor IFNGR1 to affect gene regulation IFN-gamma Protein Sheep (His) is the recombinant sheep-derived IFN-gamma protein expressed by E coli with N-6 His labeled tag