Learn More
ACROBiosystems ACROBiosystems Human IFN-gamma R1 / IFNGR1 Protein, His Tag
Supplier:  ACROBiosystems IF1H52231MG
Encompass_Preferred
Recombinant Protein;1MG;Human IFN-gamma R1, His Tag (IF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human IFN-gamma, Tag Free (Cat. No. IFG-H4211) at 5 μg/mL (100 μL/well) can bind Human IFN-gamma R1, His Tag (Cat. No. IF1-H5223) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-195-6991